Skip to Content
MilliporeSigma

CH6H9A56E018

Chem Impex International, Inc.

Resveratrol

Synonym(s):

5-(4-hydroxystyryl)benzene-1,3-diol

Sign In to View Organizational & Contract Pricing.

Select a Size

Product CH6H9A56E018 is currently not available for purchase. Contact Technical Service

Skip To

Inchi Code

1S/C14H12O3/c15-12-5-3-10(4-6-12)1-2-11-7-13(16)9-14(17)8-11/h1-9,15-17H

Inchi Key

LUKBXSAWLPMMSZ-UHFFFAOYSA-N

Shipping Temperature

RT

Purity

98%

Storage temp.

Store at - 10 C

Country of Origin

US

Physical Form

Fine white powder


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library