Skip to Content
MilliporeSigma

CH6H9A56E04A

Chem Impex International, Inc.

2-O-a-D-Glucopyranosyl-L-ascorbic acid

Sign Into View Organizational & Contract Pricing

Select a Size

Product CH6H9A56E04A is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
338.27
MDL number:

Skip To

Marketplace Support
Need help or have questions about this product?
Contact Seller

mp

158 - 163 C

Iupac Name

2-((5-(1,2-dihydroxyethyl)-2,4-dihydroxyfuran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

Inchi Code

1S/C12H18O11/c13-1-3(15)9-8(19)10(11(20)22-9)23-12-7(18)6(17)5(16)4(2-14)21-12/h3-7,12-20H,1-2H2

InChI key

MYOFXGTWVTUXOD-UHFFFAOYSA-N

storage temp.

Store at 2 - 8 C

Purity

99%

Country of Origin

US

Shipping Temperature

RT

Product Link

https://www.chemimpex.com/products/32174

Melting Point

158 - 163 C

MSDS

https://www.chemimpex.com/products/32174


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library