Skip to Content
MilliporeSigma

CH6H9A56E0A2

Chem Impex International, Inc.

D-(+)-Gluconic acid d-lactone

Synonym(s):

3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-one

Sign In to View Organizational & Contract Pricing.

Select a Size


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C6H10O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-5,7-10H,1H2

Inchi Key

PHOQVHQSTUBQQK-UHFFFAOYSA-N

Shipping Temperature

RT

Purity

99.0100.5%

Storage temp.

Store at RT

Country of Origin

US

Physical Form

White to off-white powder

Melting Point

160 C (dec.)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service