Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CH6H9A56E9D5

Chem Impex International, Inc.

L-Arginine monohydrochloride, from non-animal source, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
210.66
MDL number:
Product CH6H9A56E9D5 is currently not available for purchase. Contact Technical Service

mp

226 - 230 C

Iupac Name

2-amino-5-(diaminomethyleneamino)pentanoic acid hydrochloride

Inchi Code

1S/C6H14N4O2.ClH/c7-4(5(11)12)2-1-3-10-6(8)9;/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);1H

InChI key

KWTQSFXGGICVPE-UHFFFAOYSA-N

storage temp.

Store at RT

Purity

98.5-101.0%

Country of Origin

US

Shipping Temperature

RT

Physical Form

White crystalline powder

Product Link

https://www.chemimpex.com/product/productinfo/l-arginine-monohydrochloride-non-animal-origin/26413?cid=1

Melting Point

226 - 230 C

MSDS

https://www.chemimpex.com/product/productinfo/l-arginine-monohydrochloride-non-animal-origin/26413?cid=1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library