Skip to Content
MilliporeSigma

CH6H9A56EC7D

Chem Impex International, Inc.

Permanent orange

Synonym(s):

1-[(E)-(2,4-dinitrophenyl)diazenyl]-2-naphthol

Sign In to View Organizational & Contract Pricing.

Select a Size

25 G

$224.06

Vendor SKU: 38570-25G

$224.06


Ships in 7 days - from Aldrich Partner.


Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C16H10N4O5/c21-15-8-5-10-3-1-2-4-12(10)16(15)18-17-13-7-6-11(19(22)23)9-14(13)20(24)25/h1-9,21H

Inchi Key

HBHZKFOUIUMKHV-UHFFFAOYSA-N

Purity

90%

Country of Origin

US


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service