Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

CIAH987EB258

ChemScene LLC (Preferred Partner)

(R)-SIPHOS-PE

Synonym(s):

N,N-bis[(1R)-1-phenylethyl]-10,11,12,13-tetrahydrodiindeno[7,1-de:1,7-fg][1,3,2]dioxaphosphocin-5-amine

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C33H32O2N1P1
CAS Number:
Molecular Weight:
505.6
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C33H32NO2P/c1-23(25-11-5-3-6-12-25)34(24(2)26-13-7-4-8-14-26)37-35-29-17-9-15-27-19-21-33(31(27)29)22-20-28-16-10-18-30(36-37)32(28)33/h3-18,23-24H,19-22H2,1-2H3/t23-,24-,33-/m1/s1

Inchi Key

ZXLQLIDJAURBDD-NJQVLOCASA-N

Purity

95%

Storage temp.

-20C, stored under nitrogen

Country of Origin

CN

Physical Form

Solid

Melting Point

90-96C

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service