Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

CIAH987EBBBD

ChemScene LLC (Preferred Partner)

3-Iodo-4-methoxy-1H-pyrrolo[2,3-b]pyridine

Synonym(s):

3-iodo-1H-pyrrolo[2,3-b]pyridin-4-yl methyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

100 MG

$50.00

250 MG

$83.00

500 MG

$117.00

Vendor SKU: CS-0051311-100MG

$50.00


Ships in 5 days - from Aldrich Partner.


About This Item

Linear Formula:
C8H7O1N2I1
CAS Number:
Molecular Weight:
274.06
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C8H7IN2O/c1-12-6-2-3-10-8-7(6)5(9)4-11-8/h2-4H,1H3,(H,10,11)

Inchi Key

PGVRYUWFBSFOBO-UHFFFAOYSA-N

Purity

97%

Storage temp.

4C, protect from light

Country of Origin

CN

Physical Form

Solid

Melting Point

220-221C

Looking for similar products? Visit Product Comparison Guide

hstatements

signalword

Warning


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service