Skip to Content
MilliporeSigma

CIAH987EC000

ChemScene LLC (Preferred Partner)

4-Bromo-1H-pyrrole-2-carbonitrile

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Product CIAH987EC000 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
171
MDL number:

Skip To

Marketplace Support
Need help or have questions about this product?
Contact Seller

Iupac Name

4-bromo-1H-pyrrole-2-carbonitrile

Inchi Code

1S/C5H3BrN2/c6-4-1-5(2-7)8-3-4/h1,3,8H

InChI key

MDGARDUARHMMAR-UHFFFAOYSA-N

storage temp.

Store at room temperature

Purity

95%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/1221435-18-2.html

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0063849.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library