Sign In to View Organizational & Contract Pricing
Select a Size
10 G
$33.00
25 G
$63.00
Vendor SKU: CS-0143223-10G
$33.00
Ships in 5 days - from Aldrich Partner.
About This Item
Skip To
mp
845 C
Iupac Name
monosilver(I) monosilver(III) monosulfide
Inchi Code
1S/2Ag.S
InChI key
OCWNMDUAUUAHSJ-UHFFFAOYSA-N
storage temp.
RT, protect from light, stored under nitrogen
Purity
99%
Country of Origin
CN
Physical Form
Solid
Product Link
https://www.chemscene.com/21548-73-2.html
Melting Point
845 C
Signal Word
Danger
Hazard Statements
Precautionary statements
MSDS
https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0143223.pdf
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service