Skip to Content
MilliporeSigma

CIAH987EDCA4

ChemScene LLC (Preferred Partner)

Methyl 2-hydroxy-3-methylbutanoate

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$8.00
1 G
$45.00
2.5 G
$112.00

Vendor SKU: CS-0254529-100MG

$8.00


Ships in 5 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
132.16
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

bp

62-63 C at 15 mmHg

Iupac Name

methyl 2-hydroxy-3-methylbutanoate

Inchi Code

1S/C6H12O3/c1-4(2)5(7)6(8)9-3/h4-5,7H,1-3H3

InChI key

YSGBMDFJWFIEDF-UHFFFAOYSA-N

storage temp.

Store at room temperature

Purity

95%

Country of Origin

CN

Physical Form

Liquid

Product Link

https://www.chemscene.com/17417-00-4.html

Boiling point

62-63 C at 15 mmHg

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service