Skip to Content
MilliporeSigma

CIAH987EEB0B

ChemScene LLC (Preferred Partner)

7-Bromoquinolin-4-ol

Sign Into View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
224.06
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

bp

370.732C at 760 mmHg

mp

279-281C

Iupac Name

7-bromo-4(1H)-quinolinone

Inchi Code

1S/C9H6BrNO/c10-6-1-2-7-8(5-6)11-4-3-9(7)12/h1-5H,(H,11,12)

InChI key

GGCBEWNXEGDQAP-UHFFFAOYSA-N

storage temp.

Store at room temperature

Purity

96%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/82121-06-0.html

Flash Point

178.013C

Boiling point

370.732C at 760 mmHg

Melting Point

279-281C

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-W003315.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service