Skip to Content
MilliporeSigma

CIAH987EED54

ChemScene LLC (Preferred Partner)

3-Oxo-3-phenylpropanenitrile

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
145.16
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

bp

160C at 760 mmHg

mp

82-83C

Iupac Name

3-oxo-3-phenylpropanenitrile

Inchi Code

1S/C9H7NO/c10-7-6-9(11)8-4-2-1-3-5-8/h1-5H,6H2

InChI key

ZJRCIQAMTAINCB-UHFFFAOYSA-N

storage temp.

4C

Purity

97%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/614-16-4.html

Boiling point

160C at 760 mmHg

Melting Point

82-83C

Signal Word

Warning

Precautionary statements


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service