Skip to Content
MilliporeSigma

CIAH987EF19C

ChemScene LLC (Preferred Partner)

2-Bromothiophene-3-carbaldehyde

Synonym(s):

2-bromo-3-thiophenecarbaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

1 G

$15.00

5 G

$60.00

10 G

$111.00

25 G

$248.00

Vendor SKU: CS-W006466-1G

$15.00


Ships in 7 days - from Aldrich Partner.


About This Item

Linear Formula:
C5H3O1Br1S1
CAS Number:
Molecular Weight:
191.05
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C5H3BrOS/c6-5-4(3-7)1-2-8-5/h1-3H

Inchi Key

MMEGVQIGIBCTHI-UHFFFAOYSA-N

Purity

96%

Storage temp.

4C, protect from light, stored under nitrogen

Country of Origin

CN

Physical Form

<34C Solid,>34C Liquid

Boiling Point

60 C at 760 mmHg

Melting Point

34C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service