Skip to Content
MilliporeSigma

CIAH987EF377

ChemScene LLC (Preferred Partner)

(Bromomethyl)cyclohexane

Synonym(s):

(bromomethyl)cyclohexane

Sign In to View Organizational & Contract Pricing.

Select a Size

Product CIAH987EF377 is currently not available for purchase. Contact Technical Service

About This Item

Linear Formula:
C7H13Br1
CAS Number:
Molecular Weight:
177.08
MDL number:

Skip To

Inchi Code

1S/C7H13Br/c8-6-7-4-2-1-3-5-7/h7H,1-6H2

Inchi Key

UUWSLBWDFJMSFP-UHFFFAOYSA-N

Purity

95%

Storage temp.

Store at room temperature

Country of Origin

CN

Physical Form

Liquid

Boiling Point

175.6C at 760 mmHg

Flash Point

57C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library