Skip to Content
MilliporeSigma

CIAH987EF6C3

ChemScene LLC (Preferred Partner)

6-Bromo-1-hexene,98% (stabilized with MEHQ)

Synonym(s):

6-bromo-1-hexene

Sign In to View Organizational & Contract Pricing.

Select a Size

Product CIAH987EF6C3 is currently not available for purchase. Contact Technical Service

About This Item

Linear Formula:
C6H11Br1
CAS Number:
Molecular Weight:
163.06
MDL number:

Skip To

Inchi Code

1S/C6H11Br/c1-2-3-4-5-6-7/h2H,1,3-6H2

Inchi Key

RIMXEJYJXDBLIE-UHFFFAOYSA-N

Purity

98%

Storage temp.

4C

Country of Origin

CN

Physical Form

Liquid

Boiling Point

153.007 C at 760 mmHg

Melting Point

-102.35 C

Flash Point

52C

Density

1.2022

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library