Skip to Content
MilliporeSigma

CIAH987EF752

ChemScene LLC (Preferred Partner)

Protocatechualdehyde

Synonym(s):

3,4-dihydroxybenzaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

10 G

$8.00

25 G

$10.00

100 G

$23.00

250 G

$57.00

500 G

$113.00

1 KG

$212.00

Vendor SKU: CS-W009195-10G

$8.00


Ships in 5 days - from Aldrich Partner.


About This Item

Linear Formula:
C7H6O3
CAS Number:
Molecular Weight:
138.12
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C7H6O3/c8-4-5-1-2-6(9)7(10)3-5/h1-4,9-10H

Inchi Key

IBGBGRVKPALMCQ-UHFFFAOYSA-N

Purity

98%

Salt Data

Free Base

Storage temp.

4C, stored under nitrogen

Country of Origin

CN

Physical Form

Solid

Boiling Point

213.5 C at 760 mmHg

Melting Point

150-157C

Density

1.2667g/cm3

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service