Skip to Content
MilliporeSigma

CIAH987EF8EC

ChemScene LLC (Preferred Partner)

1-Bromo-3-methoxypropane

Sign In to View Organizational & Contract Pricing

Select a Size

25 G

$11.00

Vendor SKU: CS-W010945-25G

$11.00


Ships in 7 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
153.02
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

bp

103.5C at 760 mmHg

Iupac Name

3-bromopropyl methyl ether

Inchi Code

1S/C4H9BrO/c1-6-4-2-3-5/h2-4H2,1H3

InChI key

CEVMYGZHEJSOHZ-UHFFFAOYSA-N

storage temp.

Store at room temperature

Purity

97%

Country of Origin

CN

Physical Form

Liquid

Product Link

https://www.chemscene.com/36865-41-5.html

Flash Point

<60C

Boiling point

103.5C at 760 mmHg

Signal Word

Danger

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-W010945.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service