Skip to Content
MilliporeSigma

CIAH987EF9A1

ChemScene LLC (Preferred Partner)

Fmoc-Asp-OtBu

Sign Into View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
411.45
MDL number:
Marketplace Support
Need help or have questions about this product?
Contact Seller
Marketplace Support
Need help or have questions about this product?
Contact Seller

bp

617.4C at 760 mmHg

mp

90-98 C

Iupac Name

(3S)-4-tert-butoxy-3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-oxobutanoic acid

Inchi Code

1S/C23H25NO6/c1-23(2,3)30-21(27)19(12-20(25)26)24-22(28)29-13-18-16-10-6-4-8-14(16)15-9-5-7-11-17(15)18/h4-11,18-19H,12-13H2,1-3H3,(H,24,28)(H,25,26)/t19-/m0/s1

InChI key

VZXQYACYLGRQJU-IBGZPJMESA-N

storage temp.

4C

Purity

98%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/129460-09-9.html

Boiling point

617.4C at 760 mmHg

Melting Point

90-98 C

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-W011675.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library