Skip to Content
MilliporeSigma

CIAH987F0500

ChemScene LLC (Preferred Partner)

H-1,2,4-Triazole-3-carboxamide

Synonym(s):

4H-1,2,4-triazole-3-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Product CIAH987F0500 is currently not available for purchase. Contact Technical Service

About This Item

Linear Formula:
C3H4O1N4
CAS Number:
Molecular Weight:
112.09
MDL number:

Skip To

Inchi Code

1S/C3H4N4O/c4-2(8)3-5-1-6-7-3/h1H,(H2,4,8)(H,5,6,7)

Inchi Key

ZEWJFUNFEABPGL-UHFFFAOYSA-N

Purity

97%

Storage temp.

Store at room temperature

Country of Origin

CN

Physical Form

Solid

Boiling Point

393.4C at 760 mmHg

Melting Point

315C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library