Skip to Content
MilliporeSigma

CIAH987F0C47

ChemScene LLC (Preferred Partner)

5-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Product CIAH987F0C47 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
196.59
MDL number:

Skip To

Marketplace Support
Need help or have questions about this product?
Contact Seller

Iupac Name

5-chloro-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid

Inchi Code

1S/C8H5ClN2O2/c9-4-1-5-6(8(12)13)3-11-7(5)10-2-4/h1-3H,(H,10,11)(H,12,13)

InChI key

TUVOLKRSOFVJFQ-UHFFFAOYSA-N

storage temp.

Store at room temperature

Purity

97%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/1203498-99-0.html

Signal Word

Warning

Hazard Statements

Precautionary statements


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library