Skip to Content
MilliporeSigma

CIAH987F1DB9

ChemScene LLC (Preferred Partner)

5-Iodopent-1-yne

Synonym(s):

5-iodo-1-pentyne

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C5H7I1
CAS Number:
Molecular Weight:
194.02
MDL number:

Inchi Code

1S/C5H7I/c1-2-3-4-5-6/h1H,3-5H2

Inchi Key

SEHFYZRHGUPLSY-UHFFFAOYSA-N

Purity

96%

Storage temp.

-20C, protect from light

Country of Origin

CN

Physical Form

Liquid

Boiling Point

84-89 C at 43 mmHg

Melting Point

N/A

Flash Point

8C

Density

1.707+-0.06 g/cm3

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library