Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

CIAH987F216E

ChemScene LLC (Preferred Partner)

3-(Piperidin-1-yl)propan-1-amine

Synonym(s):

3-(1-piperidinyl)propylamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Product CIAH987F216E is currently not available for purchase. Contact Technical Service

About This Item

Linear Formula:
C8H18N2
CAS Number:
Molecular Weight:
142.24
MDL number:

Skip To

Inchi Code

1S/C8H18N2/c9-5-4-8-10-6-2-1-3-7-10/h1-9H2

Inchi Key

JMUCXULQKPWSTJ-UHFFFAOYSA-N

Purity

97%

Storage temp.

4C, protect from light

Country of Origin

CN

Physical Form

Liquid

Flash Point

82C

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library