Skip to Content
MilliporeSigma

CIAH987F36BD

ChemScene LLC (Preferred Partner)

2-(2-Fluorophenyl)cyclopropan-1-amine hydrochloride

Synonym(s):

2-(2-fluorophenyl)cyclopropanamine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C9H11N1Cl1F1
CAS Number:
Molecular Weight:
187.64
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C9H10FN.ClH/c10-8-4-2-1-3-6(8)7-5-9(7)11;/h1-4,7,9H,5,11H2;1H

Inchi Key

FTBWFKALQNGCQS-UHFFFAOYSA-N

Purity

96%

Salt Data

Hydrochloride

Storage temp.

4C, protect from light

Country of Origin

CN

Physical Form

Solid

hstatements

signalword

Warning


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service