Skip to Content
MilliporeSigma

CIAH987F38FE

ChemScene LLC (Preferred Partner)

(4-(4-methylpiperazine-1-carbonyl)phenyl)boronic acid hydrochloride

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
284.55
MDL number:

mp

187-189 C

Iupac Name

4-[(4-methyl-1-piperazinyl)carbonyl]phenylboronic acid hydrochloride

Inchi Code

1S/C12H17BN2O3.ClH/c1-14-6-8-15(9-7-14)12(16)10-2-4-11(5-3-10)13(17)18;/h2-5,17-18H,6-9H2,1H3;1H

InChI key

OUEWYSQRULKJEQ-UHFFFAOYSA-N

storage temp.

Store at room temperature

Salt Data

Hydrochloride

Purity

97%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/913835-43-5.html

Melting Point

187-189 C

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library