bp
102-104C at 1 mmHg
mp
21C
Iupac Name
3-chlorobenzenesulfonyl chloride
Inchi Code
1S/C6H4Cl2O2S/c7-5-2-1-3-6(4-5)11(8,9)10/h1-4H
InChI key
OINWZUJVEXUHCC-UHFFFAOYSA-N
storage temp.
4C
Purity
98%
Country of Origin
CN
Physical Form
Liquid
Product Link
https://www.chemscene.com/2888-06-4.html
Flash Point
110C
Boiling point
102-104C at 1 mmHg
Melting Point
21C
Signal Word
Danger
Hazard Statements
Precautionary statements
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service