Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day through March 27.Save Now

CIAH98ABF2CD

ChemScene LLC (Preferred Partner)

ANS (ammonium)

Synonym(s):

ammonium 8-anilino-1-naphthalenesulfonate

Sign In to View Organizational & Contract Pricing.

Select a Size


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C16H13NO3S.H3N/c18-21(19,20)15-11-5-7-12-6-4-10-14(16(12)15)17-13-8-2-1-3-9-13;/h1-11,17H,(H,18,19,20);1H3

Inchi Key

IPBNQYLKHUNLQE-UHFFFAOYSA-N

Purity

98%

Salt Data

Ammonium

Storage temp.

4C, sealed storage, away from moisture and light

Country of Origin

CN

Physical Form

Solid

Melting Point

242-244 C

hstatements

signalword

Warning


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service