Skip to Content
MilliporeSigma

CIAH98B92BC9

ChemScene LLC (Preferred Partner)

3-Cyano-2-phenylpropanoic acid

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Product CIAH98B92BC9 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
175.19
MDL number:

Skip To

Marketplace Support
Need help or have questions about this product?
Contact Seller

bp

365.4+-30.0 C at 760 mmHg

mp

97 C

Iupac Name

3-cyano-2-phenylpropanoic acid

Inchi Code

1S/C10H9NO2/c11-7-6-9(10(12)13)8-4-2-1-3-5-8/h1-5,9H,6H2,(H,12,13)

InChI key

XBZQKMPPVQSLHI-UHFFFAOYSA-N

storage temp.

4C

Purity

95%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/442542-97-4.html

Boiling point

365.4+-30.0 C at 760 mmHg

Melting Point

97 C

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0197761.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library