Skip to Content
MilliporeSigma

CIAH98D1E870

ChemScene LLC (Preferred Partner)

Dmt-2'-f-dc(ac) amidite

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$67.00
5 G
$276.00

Vendor SKU: CS-0149469-1G

$67.00


Ships in 5 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
789.84
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Iupac Name

(2R,5R)-5-(4-acetamido-2-oxopyrimidin-1(2H)-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-fluorotetrahydrofuran-3-yl 2-cyanoethyl diisopropylphosphoramidite

Inchi Code

1S/C41H49FN5O8P/c1-27(2)47(28(3)4)56(53-25-11-23-43)55-38-35(54-39(37(38)42)46-24-22-36(44-29(5)48)45-40(46)49)26-52-41(30-12-9-8-10-13-30,31-14-18-33(50-6)19-15-31)32-16-20-34(51-7)21-17-32/h8-10,12-22,24,27-28,35,37-39H,11,25-26H2,1-7H3,(H,44,45,48,49)/t35-,37?,38?,39-,56?/m1/s1

InChI key

CNFKJHKDSRXNFL-ARJVMWJPSA-N

storage temp.

-20C, sealed storage, away from moisture and light

Purity

95%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/159414-99-0.html

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0149469.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service