Skip to Content
MilliporeSigma

CIAH98D1F857

ChemScene LLC (Preferred Partner)

2-(Trifluoromethyl)quinolin-4-ol

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$59.00

Vendor SKU: CS-W002445-25G

$59.00


Ships in 7 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
213.16
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

bp

220.8C at 760 mmHg

mp

209-211C

Iupac Name

2-(trifluoromethyl)-4-quinolinol

Inchi Code

1S/C10H6F3NO/c11-10(12,13)9-5-8(15)6-3-1-2-4-7(6)14-9/h1-5H,(H,14,15)

InChI key

SUNAMHNJYSQUPL-UHFFFAOYSA-N

storage temp.

Store at room temperature

Purity

97%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/1701-18-4.html

Boiling point

220.8C at 760 mmHg

Melting Point

209-211C

pictograms

Signal Word

Danger

Hazard Statements


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service