Skip to Content
MilliporeSigma

CIAH98DE3760

ChemScene LLC (Preferred Partner)

3-Nitropropanol

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Product CIAH98DE3760 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
105.09
MDL number:

Skip To

Marketplace Support
Need help or have questions about this product?
Contact Seller

bp

240 C at 760 mmHg

mp

-20 C

Iupac Name

3-nitro-1-propanol

Inchi Code

1S/C3H7NO3/c5-3-1-2-4(6)7/h5H,1-3H2

InChI key

YISPKQZWSIMXMX-UHFFFAOYSA-N

storage temp.

4C

Purity

95%

Country of Origin

CN

Physical Form

Liquid

Product Link

https://www.chemscene.com/25182-84-7.html

Flash Point

116C

Boiling point

240 C at 760 mmHg

Melting Point

-20 C

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0209456.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library