Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

CIAH996D34D7

ChemScene LLC (Preferred Partner)

Ethyl 3-hydrazinylbenzoate hydrochloride

Synonym(s):

ethyl 3-hydrazinobenzoate hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Product CIAH996D34D7 is currently not available for purchase. Contact Technical Service

About This Item

Linear Formula:
C9H13O2N2Cl1
CAS Number:
Molecular Weight:
216.67
MDL number:

Skip To

Inchi Code

1S/C9H12N2O2.ClH/c1-2-13-9(12)7-4-3-5-8(6-7)11-10;/h3-6,11H,2,10H2,1H3;1H

Inchi Key

YMKGDNLSBACMSE-UHFFFAOYSA-N

Purity

98%

Salt Data

Hydrochloride

Storage temp.

4C, stored under nitrogen

Country of Origin

CN

Physical Form

Solid

Looking for similar products? Visit Product Comparison Guide

signalword

Warning

hstatements


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library