Skip to Content
MilliporeSigma

CIAH999A8170

ChemScene LLC (Preferred Partner)

Tris(2,2-bipyridyl)ruthenium(II) chloride hexahydrate

Synonym(s):

λ²-ruthenium(2+) tris(2,2'-bipyridine) methane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Storage temp.

RT, protect from light, stored under nitrogen

Inchi Code

1S/3C10H8N2.CH4.Ru/c3*1-3-7-11-9(5-1)10-6-2-4-8-12-10;;/h3*1-8H;1H4;/q;;;;+2

Inchi Key

SLZYKLALISUEFN-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Solid

Melting Point

>300C



hstatements

signalword

Warning



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library