Skip to Content
MilliporeSigma

CIAH999A8387

ChemScene LLC (Preferred Partner)

trans-1,2-Cyclohexanediamine

Synonym(s):

(1R,2R)-1,2-cyclohexanediamine

Sign In to View Organizational & Contract Pricing.

Select a Size

5 G

$12.00

10 G

$15.00

25 G

$18.00

Vendor SKU: CS-0019667-5G

$12.00


Ships in 5 days - from Aldrich Partner.


About This Item

Linear Formula:
C6H14N2
CAS Number:
Molecular Weight:
114.19
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C6H14N2/c7-5-3-1-2-4-6(5)8/h5-6H,1-4,7-8H2/t5-,6-/m1/s1

Inchi Key

SSJXIUAHEKJCMH-PHDIDXHHSA-N

Purity

97%

Storage temp.

4C, protect from light, stored under nitrogen

Country of Origin

CN

Physical Form

<14C Solid,>15C Liquid

Boiling Point

79-81C at 15 mmHg

Melting Point

14-15C

Flash Point

69C

Density

0.951g/cm3

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service