Skip to Content
MilliporeSigma

CIAH999A8687

ChemScene LLC (Preferred Partner)

Bromophenol blue (sodium)

Synonym(s):

sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)phenolate

Sign In to View Organizational & Contract Pricing.

Select a Size

Product CIAH999A8687 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
691.95
MDL number:

Skip To

mp

300 C

Inchi Code

1S/C19H10Br4O5S.Na/c20-12-5-9(6-13(21)17(12)24)19(10-7-14(22)18(25)15(23)8-10)11-3-1-2-4-16(11)29(26,27)28-19;/h1-8,24-25H;/q;+1/p-1

Inchi Key

QDVHDLHGPSKICA-UHFFFAOYSA-M

Storage temp.

Store at room temperature

Salt Data

Sodium

Purity

Country of Origin

CN

Physical Form

Solid

Looking for similar products? Visit Product Comparison Guide

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0128381.pdf

signalword

Warning


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library