Skip to Content
MilliporeSigma

CIAH99E91406

ChemScene LLC (Preferred Partner)

N-(tert-Butyl)benzenesulfonamide

Synonym(s):

N-(tert-butyl)benzenesulfonamide

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

CAS Number:
Molecular Weight:
213.3
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

bp

313.6+-25.0 C at 760 mmHg

mp

77-78 C

Inchi Code

1S/C10H15NO2S/c1-10(2,3)11-14(12,13)9-7-5-4-6-8-9/h4-8,11H,1-3H3

Inchi Key

FFUBXANSXRGVKW-UHFFFAOYSA-N

Storage temp.

Store at room temperature

Purity

96%

Country of Origin

CN

Physical Form

Solid

hstatements

signalword

Warning


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service