Inchi Code
1S/C18H33P.C13H11N.CH4O3S.Pd/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-14-13-10-6-5-9-12(13)11-7-3-2-4-8-11;1-5(2,3)4;/h16-18H,1-15H2;2-7,9-10H,1H3;1H3,(H,2,3,4);/q;-1;;+1
Inchi Key
HXNVSYATBNHKEK-UHFFFAOYSA-N
Purity
98%
Storage temp.
4C, protect from light
Country of Origin
CN
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service