Skip to Content
MilliporeSigma

CIAH9A70A794

ChemScene LLC (Preferred Partner)

6-Methylhepta-3,5-dien-2-one

Sign Into View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
124.18
MDL number:

bp

95-96 C at 20 mmHg

Iupac Name

(3E)-6-methyl-3,5-heptadien-2-one

Inchi Code

1S/C8H12O/c1-7(2)5-4-6-8(3)9/h4-6H,1-3H3/b6-4+

InChI key

KSKXSFZGARKWOW-GQCTYLIASA-N

storage temp.

4C

Purity

95%

Country of Origin

CN

Physical Form

Liquid

Product Link

https://www.chemscene.com/1604-28-0.html

Boiling point

95-96 C at 20 mmHg

Signal Word

Warning

Hazard Statements


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library