Skip to Content
MilliporeSigma

CIAH9A70A856

ChemScene LLC (Preferred Partner)

Tributylammonium (hemipyrophosphate)

Synonym(s):

tributylamine diphosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H31O7N1P2
CAS Number:
Molecular Weight:
363.33
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H27N.H4O7P2/c1-4-7-10-13(11-8-5-2)12-9-6-3;1-8(2,3)7-9(4,5)6/h4-12H2,1-3H3;(H2,1,2,3)(H2,4,5,6)

Inchi Key

WJFREWBMJPXLBS-UHFFFAOYSA-N

Purity

98%

Salt Data

pyrophosphate

Storage temp.

4C, sealed storage, away from moisture and light

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library