Skip to Content
MilliporeSigma

CIAH9A8E731A

ChemScene LLC (Preferred Partner)

7-Chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

About This Item

CAS Number:
Molecular Weight:
269.66
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Product CIAH9A8E731A is currently not available for purchase. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

bp

437.9C at 760 mmHg

mp

279-281C

Iupac Name

7-chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydro-3-quinolinecarboxylic acid

Inchi Code

1S/C12H9ClFNO3/c1-2-15-5-7(12(17)18)11(16)6-3-9(14)8(13)4-10(6)15/h3-5H,2H2,1H3,(H,17,18)

InChI key

WNNSMMJBBOPPOT-UHFFFAOYSA-N

storage temp.

Store at room temperature

Purity

97%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/68077-26-9.html

Boiling point

437.9C at 760 mmHg

Melting Point

279-281C

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0036496.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library