Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

CIAH9A8E73A3

ChemScene LLC (Preferred Partner)

N-(4-Bromo-2-nitrophenyl)acetamide

Synonym(s):

N-(4-bromo-2-nitrophenyl)acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Product CIAH9A8E73A3 is currently not available for purchase. Contact Technical Service

About This Item

Linear Formula:
C8H7O3N2Br1
CAS Number:
Molecular Weight:
259.06
MDL number:

Skip To

Inchi Code

1S/C8H7BrN2O3/c1-5(12)10-7-3-2-6(9)4-8(7)11(13)14/h2-4H,1H3,(H,10,12)

Inchi Key

GUBNCRISSRANNO-UHFFFAOYSA-N

Purity

98%

Storage temp.

Store at room temperature

Country of Origin

CN

Physical Form

Solid

Boiling Point

416.8+-35.0 C at 760 mmHg

Melting Point

102-104 C

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library