Skip to Content
MilliporeSigma

CIAH9A994799

ChemScene LLC (Preferred Partner)

Boc-Arg(di-Z)-OH

Sign Into View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
542.59
MDL number:

mp

140C

Iupac Name

(2S)-5-{[(benzyloxy)carbonyl][{[(benzyloxy)carbonyl]amino}(imino)methyl]amino}-2-[(tert-butoxycarbonyl)amino]pentanoic acid

Inchi Code

1S/C27H34N4O8/c1-27(2,3)39-25(35)29-21(22(32)33)15-10-16-31(26(36)38-18-20-13-8-5-9-14-20)23(28)30-24(34)37-17-19-11-6-4-7-12-19/h4-9,11-14,21H,10,15-18H2,1-3H3,(H,29,35)(H,32,33)(H2,28,30,34)/t21-/m0/s1

InChI key

FVWNVCDDYWITMG-NRFANRHFSA-N

storage temp.

4C

Purity

98%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/51219-19-3.html

Melting Point

140C

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library