Skip to Content
MilliporeSigma

CIAH9ABED59A

ChemScene LLC (Preferred Partner)

Dimethyl 2,6-dibromoheptanedioate

Synonym(s):

dimethyl 2,6-dibromoheptanedioate

Sign In to View Organizational & Contract Pricing.

Select a Size

Product CIAH9ABED59A is currently not available for purchase. Contact Technical Service

About This Item

Linear Formula:
C9H14O4Br2
CAS Number:
Molecular Weight:
346.02
MDL number:

Skip To

Inchi Code

1S/C9H14Br2O4/c1-14-8(12)6(10)4-3-5-7(11)9(13)15-2/h6-7H,3-5H2,1-2H3

Inchi Key

AWWJYEJSCIDADZ-UHFFFAOYSA-N

Purity

97%

Storage temp.

4C

Country of Origin

CN

Physical Form

Liquid

Boiling Point

130-140 C at 0.01 mmHg

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library