Skip to Content
MilliporeSigma

CIAH9BC7D723

ChemScene LLC (Preferred Partner)

3-Butyl-1-methyl-1H-imidazol-3-ium tetrafluoroborate

Synonym(s):

3-butyl-1-methyl-1H-imidazol-3-ium tetrafluoroborate

Sign In to View Organizational & Contract Pricing.

Select a Size

100 MG

$5.00

5 G

$10.00

10 G

$13.00

25 G

$19.00

100 G

$57.00

Vendor SKU: CS-W019912-100MG

$5.00


Ships in 7 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
226.03
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

mp

-71C

Inchi Code

1S/C8H15N2.BF4/c1-3-4-5-10-7-6-9(2)8-10;2-1(3,4)5/h6-8H,3-5H2,1-2H3;/q+1;-1

Inchi Key

LSBXQLQATZTAPE-UHFFFAOYSA-N

Storage temp.

Store at room temperature

Salt Data

Tetrafluoroborate

Purity

95%

Country of Origin

CN

Physical Form

Liquid

Flash Point

288C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service