MilliporeSigma
All Photos(1)

COM448615983

Combi-Blocks, Inc.

3-Bromo-2-(3'-bromobenzyloxy)-5-methylphenylboronic acid

CAS Number:
Molecular Weight:
399.87
MDL number:

Iupac Name

3-bromo-2-[(3-bromobenzyl)oxy]-5-methylphenylboronic acid

Inchi Code

1S/C14H13BBr2O3/c1-9-5-12(15(18)19)14(13(17)6-9)20-8-10-3-2-4-11(16)7-10/h2-7,18-19H,8H2,1H3

InChI key

VWXCNKUTFMOYDY-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Product Link

http://www.combi-blocks.com/cgi-bin/find.cgi?BB-8219

MSDS

http://www.combi-blocks.com/cgi-bin/find.cgi?BB-8219

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service