Skip to Content
MilliporeSigma

COM448640086

Combi-Blocks, Inc.

9H-Hexadecafluorononanoic acid

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$39.00
5 G
$123.50
10 G
$234.00

Vendor SKU: QA-4861-1G

$39.00


Ships in 7 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
446.09
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Iupac Name

2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluorononanoic acid

Inchi Code

1S/C9H2F16O2/c10-1(11)3(12,13)5(16,17)7(20,21)9(24,25)8(22,23)6(18,19)4(14,15)2(26)27/h1H,(H,26,27)

InChI key

RARQGXBOCXOJPM-UHFFFAOYSA-N

Purity

90%

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?QA-4861

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QA-4861


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service