Skip to Content
MilliporeSigma

COM448648731

Combi-Blocks, Inc.

X-Beta-d-glcua na 3h2o

Synonym(s):

sodium (2S,3S,4S,5R,6S)-6-((5-bromo-4-chloro-1H-indol-3-yl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylate trihydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Product COM448648731 is currently not available for purchase. Contact Technical Service

About This Item

Linear Formula:
C14H18O10N1Cl1Br1Na1
CAS Number:
Molecular Weight:
498.64
MDL number:

Skip To

Inchi Code

1S/C14H13BrClNO7.Na.3H2O/c15-4-1-2-5-7(8(4)16)6(3-17-5)23-14-11(20)9(18)10(19)12(24-14)13(21)22;;;;/h1-3,9-12,14,17-20H,(H,21,22);;3*1H2/q;+1;;;/p-1/t9-,10-,11+,12-,14+;;;;/m0..../s1

Inchi Key

KSEGXDRRKCCCGG-BTCNKGGSSA-M

Purity

95%

Storage temp.

Refrigerated

Country of Origin

CN


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library