Skip to Content
MilliporeSigma

COM448650802

Combi-Blocks, Inc.

Sodium taurocholate dihydrate

Synonym(s):

sodium 2-((4R)-4-((3R,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulfonate dihydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Product COM448650802 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
573.72
MDL number:

Skip To

Inchi Code

1S/C26H45NO7S.Na.2H2O/c1-15(4-7-23(31)27-10-11-35(32,33)34)18-5-6-19-24-20(14-22(30)26(18,19)3)25(2)9-8-17(28)12-16(25)13-21(24)29;;;/h15-22,24,28-30H,4-14H2,1-3H3,(H,27,31)(H,32,33,34);;2*1H2/q;+1;;/p-1/t15-,16?,17-,18-,19+,20+,21-,22+,24+,25+,26-;;;/m1.../s1

Inchi Key

GYRLGILDCKQBJF-SSGBDSDUSA-M

Purity

96%

Country of Origin

CN

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QB-6959


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library