MilliporeSigma
  • COM448666641
All Photos(1)

COM448666641

Combi-Blocks, Inc.

(+)-3,9-Dibromocamphor

CAS Number:
Molecular Weight:
310.03
MDL number:

Iupac Name

(1R,3S,4S)-3-bromo-7-(bromomethyl)-1,7-dimethylbicyclo[2.2.1]heptan-2-one

Inchi Code

1S/C10H14Br2O/c1-9-4-3-6(7(12)8(9)13)10(9,2)5-11/h6-7H,3-5H2,1-2H3/t6-,7+,9+,10?/m1/s1

InChI key

DCDNKSJBRIJYEC-MZIHZIJQSA-N

storage temp.

Refrigerated

Purity

97%

Country of Origin

CN

Product Link

http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2495

MSDS

http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2495

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service