COM448667227

Combi-Blocks, Inc.

N,N'-Dimethylquinacridone

CAS Number:
Molecular Weight:
340.38
MDL number:

Iupac Name

5,12-dimethyl-5,12-dihydroquino[2,3-b]acridine-7,14-dione

Inchi Code

1S/C22H16N2O2/c1-23-17-9-5-3-7-13(17)21(25)15-12-20-16(11-19(15)23)22(26)14-8-4-6-10-18(14)24(20)2/h3-12H,1-2H3

InChI key

SCZWJXTUYYSKGF-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Product Link

http://www.combi-blocks.com/cgi-bin/find.cgi?QF-8972

MSDS

http://www.combi-blocks.com/cgi-bin/find.cgi?QF-8972

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service