Documents

COM497524840

Combi-Blocks, Inc.

Z-Gln-onp

CAS Number:
Molecular Weight:
401.38
MDL number:

Iupac Name

4-nitrophenyl (2S)-5-amino-2-{[(benzyloxy)carbonyl]amino}-5-oxopentanoate

Inchi Code

1S/C19H19N3O7/c20-17(23)11-10-16(21-19(25)28-12-13-4-2-1-3-5-13)18(24)29-15-8-6-14(7-9-15)22(26)27/h1-9,16H,10-12H2,(H2,20,23)(H,21,25)/t16-/m0/s1

InChI key

KIVQPDPRDVIJDJ-INIZCTEOSA-N

Purity

95%

Country of Origin

CN

Product Link

http://www.combi-blocks.com/cgi-bin/find.cgi?QM-5242

MSDS

http://www.combi-blocks.com/cgi-bin/find.cgi?QM-5242

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.